Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-1,3,5-trimethyl-1H-pyrazole, 95%

$130.69 - $130.69

Chemical Identifiers

Chemical Name or Material 4-Bromo-1,3,5-trimethyl-1H-pyrazole
CAS 15801-69-1
Molecular Formula C6H9BrN2
Formula Weight 189.06
MDL Number MFCD00052532
Beilstein 2371
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1187606

 
alfa aesar™
L1187606
5g Each for $130.69
Description & Specifications

Specifications

Formula Weight 189.06
Chemical Name or Material 4-Bromo-1,3,5-trimethyl-1H-pyrazole
Assay 95%
Molecular Formula C6H9BrN2
MDL Number MFCD00052532
CAS 15801-69-1
Melting Point 32° to 37°C
Beilstein 2371
Boiling Point 101° to 102°C (15mmHg)
Quantity 5g