missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Bromo-1,2-difluorobenzene, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1193406

 View more versions of this product

Catalog No. AAA1193406



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Bromo-1,2-difluorobenzene
-4°C
150°C to 151°C
C6H3BrF2
MFCD00000304
UN1993
1-bromo-3,4-difluorobenzene, 3,4-difluorobromobenzene, benzene, 4-bromo-1,2-difluoro, 3,4-difluorobromebenzene, 4-bromo-1,2-difluoro-benzene, 3,4-difluoro-1-bromobenzene, paragos 390209, fr bf de, pubchem2303, 3,4-difluorophenylbromide
FC1=CC=C(Br)C=C1F
192.99
192.99
348-61-8
1.707
33°C (91°F)
1.505
5 g
1934811
YMQPKONILWWJQG-UHFFFAOYSA-N
4-bromo-1,2-difluorobenzene
67675
≥98%

Chemical Identifiers

348-61-8
192.99
YMQPKONILWWJQG-UHFFFAOYSA-N
67675
FC1=CC=C(Br)C=C1F
C6H3BrF2
MFCD00000304
1-bromo-3,4-difluorobenzene, 3,4-difluorobromobenzene, benzene, 4-bromo-1,2-difluoro, 3,4-difluorobromebenzene, 4-bromo-1,2-difluoro-benzene, 3,4-difluoro-1-bromobenzene, paragos 390209, fr bf de, pubchem2303, 3,4-difluorophenylbromide
4-bromo-1,2-difluorobenzene
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c

H226-H315-H319-H335

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

EINECSNumber : 206-481-5

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only