missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Bromo-1,1,2-trifluoro-1-butene, 98%

Manufacturer:  Alfa Aesar™ L1276609

 View more versions of this product

Catalog No. AAL1276609



Description

Description

Applications
4-Bromo-1,1,2-trifluoro-1-butene is used in organic synthesis, chemical research and pharmaceuticals.

Solubility
Not miscible or difficult to mix in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage.
Specifications

Specifications

4-Bromo-1,1,2-trifluoro-1-butene
Recognizable
10g
C4H4BrF3
MFCD00039274
1840755
GQCQMFYIFUDARF-UHFFFAOYSA-N
4-bromo-1,1,2-trifluorobut-1-ene
66333
98%
17°C (62°F)
95°C to 98°C
10493-44-4
BrCH2CH2CF=CF2
UN1993
4-bromo-1,1,2-trifluoro-1-butene, 4-bromo-1,1,2-trifluorobutene-1, 1-butene, 4-bromo-1,1,2-trifluoro, 1,1,2-trifluoro-4-bromobutene, acmc-20amo8, timtec-bb sbb006604, gqcqmfyifudarf-uhfffaoysa, 4-bromo-1,1,2-trifluorobutene., 3,4,4-trifluoro-3-butenyl bromide, 3,4,4-trifluorobut-3-enyl bromide
C(CBr)C(=C(F)F)F
188.975
188.98

Chemical Identifiers

10493-44-4
188.975
GQCQMFYIFUDARF-UHFFFAOYSA-N
66333
C(CBr)C(=C(F)F)F
C4H4BrF3
MFCD00039274
4-bromo-1,1,2-trifluoro-1-butene, 4-bromo-1,1,2-trifluorobutene-1, 1-butene, 4-bromo-1,1,2-trifluoro, 1,1,2-trifluoro-4-bromobutene, acmc-20amo8, timtec-bb sbb006604, gqcqmfyifudarf-uhfffaoysa, 4-bromo-1,1,2-trifluorobutene., 3,4,4-trifluoro-3-butenyl bromide, 3,4,4-trifluorobut-3-enyl bromide
4-bromo-1,1,2-trifluorobut-1-ene
Safety and Handling

Safety and Handling

GHS H Statement
H225-H319-H335
Highly flammable liquid and vapor.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P243-P305+P351+P338
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Take precautionary measures against static discharge.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.

Documents