Please login to your online account to display your discounted pricing

Alfa Aesar™ (+/-)-4-Boc-piperazine-2-carboxylic acid hydrate, 95%

$104.27 - $378.20

Chemical Identifiers

Chemical Name or Material (+/-)-4-Boc-piperazine-2-carboxylic acid hydrate
CAS 128019-59-0
Molecular Formula C10H18N2O4·xH2O
Formula Weight 230.26 (Anhydrous)
MDL Number MFCD01076218
Synonym 1-tert-Butyl (+/-)-piperazine-1,3-dicarboxylate; (+/-)-Piperazine-1,3-dicarboxylic acid 1-tert-butyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2741303

  SDS
alfa aesar™
H2741303
1g Each for $104.27

AAH2741306

  SDS
alfa aesar™
H2741306
5g Each for $378.20
Description & Specifications

Specifications

Formula Weight 230.26 (Anhydrous)
Quantity 1g
Chemical Name or Material (+/-)-4-Boc-piperazine-2-carboxylic acid hydrate
CAS 128019-59-0
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay 95%
MDL Number MFCD01076218
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Synonym 1-tert-Butyl (+/-)-piperazine-1,3-dicarboxylate; (+/-)-Piperazine-1,3-dicarboxylic acid 1-tert-butyl ester
Molecular Formula C10H18N2O4·xH2O

Applications
As active pharmaceutical drugs and intermediates possess different biological activities.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.