missing translation for 'onlineSavingsMsg'
Learn More

4-(Boc-aminomethyl)-N-Fmoc-L-phenylalanine, 95%

Catalog No. AAH52008MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH52008MD 250 mg
AAH5200803 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH52008MD Supplier Thermo Scientific Chemicals Supplier No. H52008MD
Only null left

CAS: 204715-91-3 | C30H32N2O6 | 516.594 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 204715-91-3
Molecular Formula C30H32N2O6
Molecular Weight (g/mol) 516.594
MDL Number MFCD01317015
InChI Key QSPDPKLGXGMDAY-SANMLTNESA-N
Synonym fmoc-4-boc-aminomethyl-l-phenylalanine, fmoc-l-4-aminomethylphe boc, 2s-3-4-tert-butoxy carbonyl amino methyl phenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-4-tert-butoxycarbonyl amino methyl phenyl propanoic acid, 2s-3-4-tert-butoxycarbonyl amino methyl phenyl-2-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, 2r-3-4-tert-butoxycarbonyl amino methyl phenyl-2-9h-fluoren-9-yl methoxycarbonyl amino propanoic acid, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino-3-4-2-methylpropan-2-yl oxycarbonylamino methyl phenyl propanoic acid, s-3-4-tert-butoxycarbonylamino-methyl-phenyl-2-9h-fluoren-9-ylmethoxycarbonylamino-propionic acid, 4-1,1-dimethylethoxy carbonyl amino methyl-n-9h-fluoren-9-ylmethoxy carbonyl-l-phenylalanine, l-phenylalanine,4-1,1-dimethylethoxy carbonyl amino methyl-n-9h-fluoren-9-ylmethoxy carbonyl
PubChem CID 2761481
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[4-[[(2-methylpropan-2-yl)oxycarbonylamino]methyl]phenyl]propanoic acid
SMILES CC(C)(C)OC(=O)NCC1=CC=C(C=C1)CC(C(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Quantity 250 mg
Formula Weight 516.59
Percent Purity 95%
Chemical Name or Material 4-(Boc-aminomethyl)-N-Fmoc-L-phenylalanine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only