Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Boc-amino)pyridine, 95%

$112.39 - $373.68

Chemical Identifiers

Chemical Name or Material 4-(Boc-amino)pyridine
CAS 98400-69-2
Molecular Formula C10H14N2O2
Formula Weight 194.23
MDL Number MFCD02179232
Synonym 4-(tert-Butoxycarbonylamino)pyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH3082006

  SDS
alfa aesar™
H3082006
5g Each for $112.39

AAH3082014

  SDS
alfa aesar™
H3082014
25g Each for $373.68
Description & Specifications

Specifications

Formula Weight 194.23
Chemical Name or Material 4-(Boc-amino)pyridine
Assay 95%
Molecular Formula C10H14N2O2
Melting Point 147° to 150°C
MDL Number MFCD02179232
Quantity 5g
CAS 98400-69-2
Synonym 4-(tert-Butoxycarbonylamino)pyridine
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Safety and Handling

Warning

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.