Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Boc-amino)-N-Fmoc-D-phenylalanine, 95%

Chemical Identifiers

Chemical Name or Material 4-(Boc-amino)-N-Fmoc-D-phenylalanine
CAS 214750-77-3
Formula Weight 502.57
MDL Number MFCD00798642
Synonym Fmoc-D-Phe(4-NHBoc)-OH
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6661606

 
alfa aesar™
H6661606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6661603

 
alfa aesar™
H6661603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH66616MD

 
alfa aesar™
H66616MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 502.57
CAS 214750-77-3
MDL Number MFCD00798642
Assay 95%
Physical Form Solid
Quantity 5g
Storage Note 1 -30° to -10°C
Chemical Name or Material 4-(Boc-amino)-N-Fmoc-D-phenylalanine
Synonym Fmoc-D-Phe(4-NHBoc)-OH
Safety and Handling

-30° to -10°C