missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Boc-amino)indole-3-carboxaldehyde, 97%

$173.12 - $600.92

Chemical Identifiers

CAS 885266-77-3
Molecular Formula C14H16N2O3
Molecular Weight (g/mol) 260.293
MDL Number MFCD04973990
InChI Key XKYBSNNENLRYPN-UHFFFAOYSA-N
Synonym 3-formyl-1h-indol-4-yl-carbamic acid tert-butyl ester, 3-formyl-indol-4-yl-carbamic acid tert-butyl ester, tert-butyl 3-formyl-1h-indol-4-ylcarbamate, tert-butyl n-3-formyl-1h-indol-4-yl carbamate
PubChem CID 24720953
IUPAC Name tert-butyl N-(3-formyl-1H-indol-4-yl)carbamate
SMILES CC(C)(C)OC(=O)NC1=CC=CC2=C1C(=CN2)C=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH63587MD View Documents Alfa Aesar™
H63587MD
250mg Each for $173.12
 
AAH6358703 View Documents Alfa Aesar™
H6358703
1g Each for $600.92
 
Specifications

Chemical Identifiers

885266-77-3
260.293
XKYBSNNENLRYPN-UHFFFAOYSA-N
24720953
CC(C)(C)OC(=O)NC1=CC=CC2=C1C(=CN2)C=O
C14H16N2O3
MFCD04973990
3-formyl-1h-indol-4-yl-carbamic acid tert-butyl ester, 3-formyl-indol-4-yl-carbamic acid tert-butyl ester, tert-butyl 3-formyl-1h-indol-4-ylcarbamate, tert-butyl n-3-formyl-1h-indol-4-yl carbamate
tert-butyl N-(3-formyl-1H-indol-4-yl)carbamate

Specifications

250mg
C14H16N2O3
3-formyl-1h-indol-4-yl-carbamic acid tert-butyl ester, 3-formyl-indol-4-yl-carbamic acid tert-butyl ester, tert-butyl 3-formyl-1h-indol-4-ylcarbamate, tert-butyl n-3-formyl-1h-indol-4-yl carbamate
CC(C)(C)OC(=O)NC1=CC=CC2=C1C(=CN2)C=O
260.293
260.29
97%
885266-77-3
MFCD04973990
XKYBSNNENLRYPN-UHFFFAOYSA-N
tert-butyl N-(3-formyl-1H-indol-4-yl)carbamate
24720953
Solid
4-(Boc-amino)indole-3-carboxaldehyde
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold