missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Boc-amino)indole-3-carboxaldehyde, 97%

Manufacturer:  Alfa Aesar™ H63587MD

 View more versions of this product

Catalog No. AAH63587MD



Specifications

Specifications

4-(Boc-amino)indole-3-carboxaldehyde
885266-77-3
MFCD04973990
XKYBSNNENLRYPN-UHFFFAOYSA-N
tert-butyl N-(3-formyl-1H-indol-4-yl)carbamate
24720953
Solid
250mg
C14H16N2O3
tert-butyl 3-formyl-1h-indol-4-ylcarbamate, tert-butyl n-3-formyl-1h-indol-4-yl carbamate, 3-formyl-indol-4-yl-carbamic acid tert-butyl ester, 3-formyl-1h-indol-4-yl-carbamic acid tert-butyl ester
CC(C)(C)OC(=O)NC1=CC=CC2=C1C(=CN2)C=O
260.293
260.29
97%

Chemical Identifiers

885266-77-3
260.293
XKYBSNNENLRYPN-UHFFFAOYSA-N
24720953
CC(C)(C)OC(=O)NC1=CC=CC2=C1C(=CN2)C=O
C14H16N2O3
MFCD04973990
tert-butyl 3-formyl-1h-indol-4-ylcarbamate, tert-butyl n-3-formyl-1h-indol-4-yl carbamate, 3-formyl-indol-4-yl-carbamic acid tert-butyl ester, 3-formyl-1h-indol-4-yl-carbamic acid tert-butyl ester
tert-butyl N-(3-formyl-1H-indol-4-yl)carbamate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

storageNote1 : 2° to 8°C

Documents