Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-(Boc-amino)biphenyl-3-carboxylic acid, 95%

Chemical Identifiers

Chemical Name or Material 4'-(Boc-amino)biphenyl-3-carboxylic acid
Molecular Formula C18H19NO4
Formula Weight 313.35
MDL Number MFCD09055096
Synonym 4'-(tert-Butoxycarbonylamino)biphenyl-3-carboxylic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5191803

  SDS
alfa aesar™
H5191803
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5191806

  SDS
alfa aesar™
H5191806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 313.35
Chemical Name or Material 4'-(Boc-amino)biphenyl-3-carboxylic acid
Assay 95%
Molecular Formula C18H19NO4
MDL Number MFCD09055096
Quantity 1g
Synonym 4'-(tert-Butoxycarbonylamino)biphenyl-3-carboxylic acid