missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-(Boc-amino)benzonitrile, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals H54272MD

 View more versions of this product

Catalog No. AAH54272MD



Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

4-(Boc-amino)benzonitrile
C12H14N2O2
250 mg
tert-butyl 4-cyanophenylcarbamate, tert-butyl 4-cyanophenyl carbamate, 4-boc-amino benzonitrile, tert-butyl n-4-cyanophenyl carbamate, carbamic acid,n-4-cyanophenyl-, 1,1-dimethylethyl ester, acmc-1c548, n-boc-4-aminobenzonitrile, tert-butyl 4-cyanophenyl-carbamate, 4-cyano-tert-butyloxycarbonylaniline
CC(C)(C)OC(=O)NC1=CC=C(C=C1)C#N
218.256
218.26
143090-18-0
MFCD08703141
6326835
CPIRTCVNDVQQLE-UHFFFAOYSA-N
tert-butyl N-(4-cyanophenyl)carbamate
10130644
97%

Chemical Identifiers

143090-18-0
218.256
CPIRTCVNDVQQLE-UHFFFAOYSA-N
10130644
CC(C)(C)OC(=O)NC1=CC=C(C=C1)C#N
C12H14N2O2
MFCD08703141
tert-butyl 4-cyanophenylcarbamate, tert-butyl 4-cyanophenyl carbamate, 4-boc-amino benzonitrile, tert-butyl n-4-cyanophenyl carbamate, carbamic acid,n-4-cyanophenyl-, 1,1-dimethylethyl ester, acmc-1c548, n-boc-4-aminobenzonitrile, tert-butyl 4-cyanophenyl-carbamate, 4-cyano-tert-butyloxycarbonylaniline
tert-butyl N-(4-cyanophenyl)carbamate
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions