missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Boc-amino)benzonitrile, 97%

Manufacturer:  Alfa Aesar™ H5427206

 View more versions of this product

Catalog No. AAH5427206



Specifications

Specifications

4-(Boc-amino)benzonitrile
143090-18-0
MFCD08703141
4-boc-amino benzonitrile
CC(C)(C)OC(=O)NC1=CC=C(C=C1)C#N
218.256
218.26
5g
C12H14N2O2
6326835
CPIRTCVNDVQQLE-UHFFFAOYSA-N
tert-butyl N-(4-cyanophenyl)carbamate
10130644
97%

Chemical Identifiers

143090-18-0
218.256
CPIRTCVNDVQQLE-UHFFFAOYSA-N
10130644
CC(C)(C)OC(=O)NC1=CC=C(C=C1)C#N
C12H14N2O2
MFCD08703141
4-boc-amino benzonitrile
tert-butyl N-(4-cyanophenyl)carbamate
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents