missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Boc-amino)benzonitrile, 97%

Manufacturer:  Alfa Aesar™ H5427203

 View more versions of this product

Catalog No. AAH5427203



Specifications

Specifications

4-(Boc-amino)benzonitrile
143090-18-0
MFCD08703141
4-boc-amino benzonitrile
CC(C)(C)OC(=O)NC1=CC=C(C=C1)C#N
218.256
218.26
1g
C12H14N2O2
6326835
CPIRTCVNDVQQLE-UHFFFAOYSA-N
tert-butyl N-(4-cyanophenyl)carbamate
10130644
97%

Chemical Identifiers

143090-18-0
218.256
CPIRTCVNDVQQLE-UHFFFAOYSA-N
10130644
CC(C)(C)OC(=O)NC1=CC=C(C=C1)C#N
C12H14N2O2
MFCD08703141
4-boc-amino benzonitrile
tert-butyl N-(4-cyanophenyl)carbamate
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Documents