Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Boc-amino)benzoic acid, 97%

$50.34 - $116.68

Chemical Identifiers

Chemical Name or Material 4-(Boc-amino)benzoic acid
CAS 66493-39-8
Molecular Formula C12H15NO4
Formula Weight 237.26
MDL Number MFCD00037428
Beilstein 2115614
Synonym N-tert-Butoxycarbonyl-4-aminobenzoic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1853103

 
alfa aesar™
L1853103
1g Each for $50.34

AAL1853106

 
alfa aesar™
L1853106
5g Each for $116.68
Description & Specifications

Specifications

Formula Weight 237.26
Chemical Name or Material 4-(Boc-amino)benzoic acid
MDL Number MFCD00037428
CAS 66493-39-8
Assay 97%
Molecular Formula C12H15NO4
Melting Point ≈193°C
Beilstein 2115614
Quantity 1g
Synonym N-tert-Butoxycarbonyl-4-aminobenzoic acid
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.