missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-(Boc-amino)-4-carbamoylbutyric acid, 98%, Thermo Scientific Chemicals
$84.53 - $1097.75
Chemical Identifiers
CAS | 18800-74-3 |
---|---|
Molecular Formula | C10H17N2O5 |
Molecular Weight (g/mol) | 245.26 |
MDL Number | MFCD00070081 |
InChI Key | GMZBDBBBJXGPIH-LURJTMIESA-M |
Synonym | boc-glu-nh2, n-alpha-tert-boc-l-isoglutamine, 4s-4-n-tert-butoxycarbonylamino-4-carbamoyl butanoic acid, 4s-4-tert-butoxycarbonyl amino-4-carbamoylbutanoic acid, boc-glu-nh, ambotzbaa1095, na-t-boc-l-isoglutamine, boc-isogln-oh;boc-l-isoglutamine, n-alpha-t-butyloxycarbonyl-l-glutamic amide, 4-boc-amino-4-carbamoylbutyric acid |
PubChem CID | 2734757 |
IUPAC Name | (4S)-5-amino-4-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid |
SMILES | CC(C)(C)OC(=O)N[C@@H](CCC([O-])=O)C(N)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAH62305MD
|
Thermo Scientific Chemicals
H62305MD |
250 mg |
Each for $84.53
|
|
|||||
AAH6230503
|
Thermo Scientific Chemicals
H6230503 |
1 g |
Each for $254.05
|
|
|||||
AAH6230506
|
Thermo Scientific Chemicals
H6230506 |
5 g |
Each for $1,097.75
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
18800-74-3 | |
245.26 | |
GMZBDBBBJXGPIH-LURJTMIESA-M | |
2734757 | |
CC(C)(C)OC(=O)N[C@@H](CCC([O-])=O)C(N)=O |
C10H17N2O5 | |
MFCD00070081 | |
boc-glu-nh2, n-alpha-tert-boc-l-isoglutamine, 4s-4-n-tert-butoxycarbonylamino-4-carbamoyl butanoic acid, 4s-4-tert-butoxycarbonyl amino-4-carbamoylbutanoic acid, boc-glu-nh, ambotzbaa1095, na-t-boc-l-isoglutamine, boc-isogln-oh;boc-l-isoglutamine, n-alpha-t-butyloxycarbonyl-l-glutamic amide, 4-boc-amino-4-carbamoylbutyric acid | |
(4S)-5-amino-4-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid |
Specifications
18800-74-3 | |
C10H17N2O5 | |
250 mg | |
GMZBDBBBJXGPIH-LURJTMIESA-M | |
(4S)-5-amino-4-[(2-methylpropan-2-yl)oxycarbonylamino]-5-oxopentanoic acid | |
2734757 | |
98% | |
4-(Boc-amino)-4-carbamoylbutyric acid |
White | |
MFCD00070081 | |
boc-glu-nh2, n-alpha-tert-boc-l-isoglutamine, 4s-4-n-tert-butoxycarbonylamino-4-carbamoyl butanoic acid, 4s-4-tert-butoxycarbonyl amino-4-carbamoylbutanoic acid, boc-glu-nh, ambotzbaa1095, na-t-boc-l-isoglutamine, boc-isogln-oh;boc-l-isoglutamine, n-alpha-t-butyloxycarbonyl-l-glutamic amide, 4-boc-amino-4-carbamoylbutyric acid | |
CC(C)(C)OC(=O)N[C@@H](CCC([O-])=O)C(N)=O | |
245.26 | |
246.26 | |
Powder |
Safety and Handling
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only