Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Boc-amino)-3-iodopyridine, 96%

$78.04 - $213.96

Chemical Identifiers

Chemical Name or Material 4-(Boc-amino)-3-iodopyridine
CAS 211029-67-3
Molecular Formula C10H13IN2O2
Formula Weight 320.13
MDL Number MFCD04973409
Synonym 4-(tert-Butoxycarbonylamino)-3-iodopyridine; tert-Butyl (3-iodo-4-pyridyl)carbamate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2875103

 
alfa aesar™
H2875103
1g Each for $213.96

AAH28751MD

 
alfa aesar™
H28751MD
250mg Each for $78.04
Description & Specifications

Specifications

Formula Weight 320.13
Chemical Name or Material 4-(Boc-amino)-3-iodopyridine
Assay 96%
Molecular Formula C10H13IN2O2
Melting Point 79° to 83°C
MDL Number MFCD04973409
Quantity 1g
CAS 211029-67-3
Synonym 4-(tert-Butoxycarbonylamino)-3-iodopyridine; tert-Butyl (3-iodo-4-pyridyl)carbamate
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.