missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Boc-amino)-2-chloropyridine, 95%

$229.82 - $1,015.76

Chemical Identifiers

CAS 234108-73-7
Molecular Formula C10H13ClN2O2
Molecular Weight (g/mol) 228.676
MDL Number MFCD02179232
InChI Key SJCKHLJWAXGPGT-UHFFFAOYSA-N
Synonym 2-chloro-pyridin-4-yl-carbamic acid tert-butyl ester
PubChem CID 21858447
IUPAC Name tert-butyl N-(2-chloropyridin-4-yl)carbamate
SMILES CC(C)(C)OC(=O)NC1=CC(=NC=C1)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5006703 View Documents Alfa Aesar™
H5006703
1g Each for $229.82
 
AAH5006706 View Documents Alfa Aesar™
H5006706
5g Each for $1,015.76
 
Specifications

Chemical Identifiers

234108-73-7
228.676
SJCKHLJWAXGPGT-UHFFFAOYSA-N
21858447
CC(C)(C)OC(=O)NC1=CC(=NC=C1)Cl
C10H13ClN2O2
MFCD02179232
2-chloro-pyridin-4-yl-carbamic acid tert-butyl ester
tert-butyl N-(2-chloropyridin-4-yl)carbamate

Specifications

1g
C10H13ClN2O2
2-chloro-pyridin-4-yl-carbamic acid tert-butyl ester
CC(C)(C)OC(=O)NC1=CC(=NC=C1)Cl
228.676
228.68
4-(Boc-amino)-2-chloropyridine
234108-73-7
MFCD02179232
SJCKHLJWAXGPGT-UHFFFAOYSA-N
tert-butyl N-(2-chloropyridin-4-yl)carbamate
21858447
95%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning