Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Boc-amino)-2-bromopyridine, 97%

$130.84 - $130.84

Chemical Identifiers

Chemical Name or Material 4-(Boc-amino)-2-bromopyridine
CAS 433711-95-6
Molecular Formula C10H13BrN2O2
Formula Weight 273.13g/mol
MDL Number MFCD03791266
Synonym 2-Bromo-4-(tert-butoxycarbonylamino)pyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1984703

 
alfa aesar™
L19847-03
1g Each for $130.84
Description & Specifications

Specifications

Quantity 1g
Chemical Name or Material 4-(Boc-amino)-2-bromopyridine
CAS 433711-95-6
Assay 97%
Molecular Formula C10H13BrN2O2
Formula Weight 273.13g/mol
MDL Number MFCD03791266
Synonym 2-Bromo-4-(tert-butoxycarbonylamino)pyridine