Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-(Boc-amino)-1-indoleacetic acid, 97%

Chemical Identifiers

Chemical Name or Material 4-(Boc-amino)-1-indoleacetic acid
CAS 885266-64-8
Formula Weight 290.31g/mol
MDL Number MFCD04973968
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6396203

 
alfa aesar™
H63962-03
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH63962MD

 
alfa aesar™
H63962-MD
250mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 1g
Chemical Name or Material 4-(Boc-amino)-1-indoleacetic acid
CAS 885266-64-8
Physical Form Solid
Assay 97%
Formula Weight 290.31g/mol
MDL Number MFCD04973968
Storage Note 1 2° to 8°C
Safety and Handling

2° to 8°C