missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Boc-1-(6-methyl-2-pyridyl)piperazine, 97%

Manufacturer:  Alfa Aesar™ H5584106

 View more versions of this product

Catalog No. AAH5584106



Specifications

Specifications

4-Boc-1-(6-methyl-2-pyridyl)piperazine
80°C to 84°C
127188-33-4
MFCD07369771
1-piperazinecarboxylicacid, 4-6-methyl-2-pyridinyl-, 1,1-dimethylethyl ester
CC1=NC(=CC=C1)N2CCN(CC2)C(=O)OC(C)(C)C
277.368
277.37
97%
White
5g
C15H23N3O2
UN2811
QEDVZUFNZJJSJL-UHFFFAOYSA-N
tert-butyl 4-(6-methylpyridin-2-yl)piperazine-1-carboxylate
16217938
Powder

Chemical Identifiers

127188-33-4
277.368
QEDVZUFNZJJSJL-UHFFFAOYSA-N
16217938
CC1=NC(=CC=C1)N2CCN(CC2)C(=O)OC(C)(C)C
C15H23N3O2
MFCD07369771
1-piperazinecarboxylicacid, 4-6-methyl-2-pyridinyl-, 1,1-dimethylethyl ester
tert-butyl 4-(6-methylpyridin-2-yl)piperazine-1-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

Documents