Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Boc-1-(6-bromo-2-pyridyl)piperazine, 97%

Chemical Identifiers

Chemical Name or Material 4-Boc-1-(6-bromo-2-pyridyl)piperazine
CAS 331767-56-7
Formula Weight 342.23
MDL Number MFCD07369773
UN Number UN2811
Synonym 2-(1-Boc-1-piperazinyl)-6-bromopyridine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5520903

 
alfa aesar™
H5520903
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5520906

 
alfa aesar™
H5520906
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5520914

 
alfa aesar™
H5520914
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 94° to 98°C
Formula Weight 342.23
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 4-Boc-1-(6-bromo-2-pyridyl)piperazine
Physical Form Powder
Assay 97%
MDL Number MFCD07369773
Quantity 1g
CAS 331767-56-7
UN Number UN2811
Color White to yellow
Synonym 2-(1-Boc-1-piperazinyl)-6-bromopyridine
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
DOT Information Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.