Learn More
4-Boc-1-(6-bromo-2-pyridyl)piperazine, 97%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals H5520903
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
4-Boc-1-(6-bromo-2-pyridyl)piperazine | |
94°C to 98°C | |
C14H20BrN3O2 | |
1 g | |
4-boc-1-6-bromo-2-pyridyl piperazine, tert-butyl 4-6-bromopyridin-2-yl piperazine-1-carboxylate, t-butyl 4-6-bromopyridin-2-yl piperazine-1-carboxylate, 2-n-boc-piperazin-1-yl-6-bromopyridine, 1-piperazinecarboxylic acid, 4-6-bromo-2-pyridinyl-, 1,1-dimethylethyl ester, acmc-1ctvy, 4-boc-1-6-bromo-pyridin-2-yl piperazine, tert-butyl 4-6'-bromopyrid-2'-yl piperazine-1-carboxylate, tert.butyl 4-6-bromo-pyridin-2-yl-piperazine-1-carboxylate | |
CC(C)(C)OC(=O)N1CCN(CC1)C2=NC(=CC=C2)Br | |
342.237 | |
342.23 | |
Powder |
331767-56-7 | |
White to Yellow | |
MFCD07369773 | |
UN2811 | |
CQSAPHUUCUUHNS-UHFFFAOYSA-N | |
tert-butyl 4-(6-bromopyridin-2-yl)piperazine-1-carboxylate | |
16059046 | |
97% |
Chemical Identifiers
331767-56-7 | |
342.237 | |
CQSAPHUUCUUHNS-UHFFFAOYSA-N | |
16059046 | |
CC(C)(C)OC(=O)N1CCN(CC1)C2=NC(=CC=C2)Br |
C14H20BrN3O2 | |
MFCD07369773 | |
4-boc-1-6-bromo-2-pyridyl piperazine, tert-butyl 4-6-bromopyridin-2-yl piperazine-1-carboxylate, t-butyl 4-6-bromopyridin-2-yl piperazine-1-carboxylate, 2-n-boc-piperazin-1-yl-6-bromopyridine, 1-piperazinecarboxylic acid, 4-6-bromo-2-pyridinyl-, 1,1-dimethylethyl ester, acmc-1ctvy, 4-boc-1-6-bromo-pyridin-2-yl piperazine, tert-butyl 4-6'-bromopyrid-2'-yl piperazine-1-carboxylate, tert.butyl 4-6-bromo-pyridin-2-yl-piperazine-1-carboxylate | |
tert-butyl 4-(6-bromopyridin-2-yl)piperazine-1-carboxylate |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H301-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
TSCA : No
Recommended Storage : Ambient temperatures