missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Boc-1-(5-bromo-2-pyridyl)piperazine, 97%

Manufacturer:  Alfa Aesar™ H5531106

 View more versions of this product

Catalog No. AAH5531106



Description

Description

Applications
4-Boc-1-(5-bromo-2-pyridyl)piperazine is used as active pharmaceutical ingredient.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

4-Boc-1-(5-bromo-2-pyridyl)piperazine
83°C to 87°C
153747-97-8
MFCD07369772
1-piperazinecarboxylic acid, 4-5-bromo-2-pyridinyl-, 1,1-dimethylethyl ester
DSLVSFMWCDGZIL-UHFFFAOYSA-N
tert-butyl 4-(5-bromopyridin-2-yl)piperazine-1-carboxylate
11244775
Powder
White to Yellow
5g
C14H20BrN3O2
UN2811
Soluble in water.
CC(C)(C)OC(=O)N1CCN(CC1)C2=NC=C(C=C2)Br
342.237
342.23
97%

Chemical Identifiers

153747-97-8
342.237
DSLVSFMWCDGZIL-UHFFFAOYSA-N
11244775
CC(C)(C)OC(=O)N1CCN(CC1)C2=NC=C(C=C2)Br
C14H20BrN3O2
MFCD07369772
1-piperazinecarboxylic acid, 4-5-bromo-2-pyridinyl-, 1,1-dimethylethyl ester
tert-butyl 4-(5-bromopyridin-2-yl)piperazine-1-carboxylate
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H301-H315-H319-H335

DOTInformation : Transport Hazard Class: 6.1; Packaging Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

TSCA : No

Recommended Storage : Ambient temperatures

Documents