Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-[Bis(2-methoxyethyl)aminomethyl]benzeneboronic acid pinacol ester, 95%

Chemical Identifiers

Chemical Name or Material 4-[Bis(2-methoxyethyl)aminomethyl]benzeneboronic acid pinacol ester
Molecular Formula C19H32BNO4
Formula Weight 349.28
MDL Number MFCD16294545
Synonym Bis(2-methoxyethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzylamine
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5194203

 
alfa aesar™
H5194203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5194206

 
alfa aesar™
H5194206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 349.28
Chemical Name or Material 4-[Bis(2-methoxyethyl)aminomethyl]benzeneboronic acid pinacol ester
Assay 95%
Molecular Formula C19H32BNO4
MDL Number MFCD16294545
Synonym Bis(2-methoxyethyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzylamine
Quantity 1g