Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-[Bis(2-hydroxyethyl)amino]benzylidenemalononitrile, 95%

$158.45 - $158.45

Chemical Identifiers

Chemical Name or Material 4-[Bis(2-hydroxyethyl)amino]benzylidenemalononitrile
CAS 63619-34-1
Molecular Formula C14H15N3O2
Formula Weight 257.29g/mol
MDL Number MFCD06200699
Synonym 4-(2,2-Dicyanovinyl)-N-bis(hydroxyethyl)aniline
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5108103

 
alfa aesar™
H51081-03
1g Each for $158.45
Description & Specifications

Specifications

Quantity 1g
Quantity 1g
Chemical Name or Material 4-[Bis(2-hydroxyethyl)amino]benzylidenemalononitrile
CAS 63619-34-1
Assay 95%
Molecular Formula C14H15N3O2
Formula Weight 257.29g/mol
MDL Number MFCD06200699
Synonym 4-(2,2-Dicyanovinyl)-N-bis(hydroxyethyl)aniline
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H317
May cause an allergic skin reaction.
Health Hazard 3 GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P363-P321-P333+P313-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
Wash contaminated clothing before reuse.
Specific treatment (see label).
If skin irritation or rash occurs: Get medical advice/attention.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H317
May cause an allergic skin reaction.