missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Biphenylyl Trifluoromethanesulfonate 98.0+%, TCI America™
$86.27 - $255.49
Chemical Identifiers
CAS | 17763-78-9 |
---|---|
Molecular Formula | C13H9F3O3S |
Molecular Weight (g/mol) | 302.267 |
MDL Number | MFCD18379753 |
InChI Key | RJFYGCJTUUXOOF-UHFFFAOYSA-N |
Synonym | Trifluoromethanesulfonic Acid 4-Biphenylyl Ester, 4-Biphenylyl Triflate |
PubChem CID | 11077493 |
IUPAC Name | (4-phenylphenyl) trifluoromethanesulfonate |
SMILES | C1=CC=C(C=C1)C2=CC=C(C=C2)OS(=O)(=O)C(F)(F)F |
Chemical Identifiers
17763-78-9 | |
302.267 | |
RJFYGCJTUUXOOF-UHFFFAOYSA-N | |
11077493 | |
C1=CC=C(C=C1)C2=CC=C(C=C2)OS(=O)(=O)C(F)(F)F |
C13H9F3O3S | |
MFCD18379753 | |
Trifluoromethanesulfonic Acid 4-Biphenylyl Ester, 4-Biphenylyl Triflate | |
(4-phenylphenyl) trifluoromethanesulfonate |
Specifications
17763-78-9 | |
White-Yellow | |
MFCD18379753 | |
Trifluoromethanesulfonic Acid 4-Biphenylyl Ester, 4-Biphenylyl Triflate | |
C1=CC=C(C=C1)C2=CC=C(C=C2)OS(=O)(=O)C(F)(F)F | |
302.267 | |
302.27 | |
Crystalline Powder |
54°C | |
C13H9F3O3S | |
1 g | |
RJFYGCJTUUXOOF-UHFFFAOYSA-N | |
(4-phenylphenyl) trifluoromethanesulfonate | |
11077493 | |
≥98.0% (GC) | |
4-Biphenylyl Trifluoromethanesulfonate |
Safety and Handling
TSCA : No