Please login to your online account to display your discounted pricing

4-Biphenylmethanol, 98%, ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC154130500

 View more versions of this product

Catalog No. AC154130500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Powder
Formula Weight 184.24
Quantity 50g
Color White to beige
Melting Point 101° to 105°C
Beilstein 06, II, 636
CAS Max % 100.0
Linear Formula C6H5C6H4CH2OH
CAS 3597-91-9
Molecular Formula C13H12O
Chemical Name or Material 4-Biphenylmethanol
CAS Min % 97.5
MDL Number MFCD00004660
Packaging Glass bottle
Synonym 4-Phenylbenzyl alcohol
Infrared Spectrum Authentic
Assay Percent Range 98%

Specifications

Chemical Name or Material 4-Biphenylmethanol
CAS 3597-91-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C13H12O
Linear Formula C6H5C6H4CH2OH
Formula Weight 184.24
MDL Number MFCD00004660
Synonym 4-Phenylbenzyl alcohol