Please login to your online account to display your discounted pricing

4-Biphenylmethanol, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Biphenylmethanol
CAS 3597-91-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C13H12O
Linear Formula C6H5C6H4CH2OH
Formula Weight 184.24
MDL Number MFCD00004660
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC154130500

  SDS
thermo fisher scientific
AC154130500
Glass bottle 50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Powder
Formula Weight 184.24
Quantity 50g
Color White to beige
Melting Point 101° to 105°C
Beilstein 06, II, 636
CAS Max % 100.0
Molecular Formula C13H12O
Linear Formula C6H5C6H4CH2OH
CAS 3597-91-9
Chemical Name or Material 4-Biphenylmethanol
Packaging Glass bottle
MDL Number MFCD00004660
Synonym 4-Phenylbenzyl alcohol
CAS Min % 97.5
Infrared Spectrum Authentic
Assay Percent Range 98%