missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Biphenylcarbonyl chloride, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 171250500

 View more versions of this product

Catalog No. AC171250500

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

4-Biphenylcarbonyl chloride
Glass bottle
110°C to 114°C
98%
97.5
97.5% min. (ex Cl) (Argentometry)
C6H5C6H4COCl
09, I, 280
1,1'-biphenyl-4-carbonyl chloride, 1,1'-biphenyl-4carbonyl chloride, 4-biphenyl carbonyl chloride, 4-biphenylcarbonyl chloride, 4-biphenylcarbonylchloride, 4-phenyl benzoyl chloride, biphenyl-4-carbonyl chloride, p-phenylbenzoyl chloride
JPVUWCPKMYXOKW-UHFFFAOYSA-N
4-phenylbenzoyl chloride
84151
Fine Crystalline Powder
Authentic
White to Yellow
50g
14002-51-8
100.0
C13H9ClO
MFCD00000692
04,376; 07,282
Solubility in water: reacts. Other solubilities: soluble in chloroform,dmf
C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)Cl
216.67
216.67
98%

Chemical Identifiers

14002-51-8
216.67
JPVUWCPKMYXOKW-UHFFFAOYSA-N
84151
C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)Cl
C13H9ClO
MFCD00000692
1,1'-biphenyl-4-carbonyl chloride, 1,1'-biphenyl-4carbonyl chloride, 4-biphenyl carbonyl chloride, 4-biphenylcarbonyl chloride, 4-biphenylcarbonylchloride, 4-phenyl benzoyl chloride, biphenyl-4-carbonyl chloride, p-phenylbenzoyl chloride
4-phenylbenzoyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
Reacts violently with water.

GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi

GHS Signal Word: Danger

EINECSNumber : 237-804-8

TSCA : TSCA

SDS Documents