Learn More
4-Biphenylacetic acid, 98%, Thermo Scientific Chemicals
$269.26 - $1262.89
Chemical Identifiers
CAS | 5728-52-9 |
---|---|
Molecular Formula | C14H12O2 |
Molecular Weight (g/mol) | 212.248 |
MDL Number | MFCD00004351 |
InChI Key | QRZAKQDHEVVFRX-UHFFFAOYSA-N |
Synonym | 4-biphenylacetic acid, felbinac, 4-biphenylylacetic acid, traxam, dolinac, 4-carboxymethylbiphenyl, napageln, 2-4-phenylphenyl acetic acid, p-biphenylylacetic acid, 1,1'-biphenyl-4-acetic acid |
PubChem CID | 3332 |
ChEBI | CHEBI:31597 |
IUPAC Name | 2-(4-phenylphenyl)acetic acid |
SMILES | C1=CC=C(C=C1)C2=CC=C(C=C2)CC(=O)O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAA1174918
|
Thermo Scientific Chemicals
A1174918 |
50 g |
Each for $269.26
|
|
|||||
AAA1174930
|
Thermo Scientific Chemicals
A1174930 |
250 g |
Each for $1,262.89
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
5728-52-9 | |
212.248 | |
QRZAKQDHEVVFRX-UHFFFAOYSA-N | |
3332 | |
2-(4-phenylphenyl)acetic acid |
C14H12O2 | |
MFCD00004351 | |
4-biphenylacetic acid, felbinac, 4-biphenylylacetic acid, traxam, dolinac, 4-carboxymethylbiphenyl, napageln, 2-4-phenylphenyl acetic acid, p-biphenylylacetic acid, 1,1'-biphenyl-4-acetic acid | |
CHEBI:31597 | |
C1=CC=C(C=C1)C2=CC=C(C=C2)CC(=O)O |
Specifications
5728-52-9 | |
Odorless | |
MFCD00004351 | |
UN2811 | |
14,3951 | |
QRZAKQDHEVVFRX-UHFFFAOYSA-N | |
2-(4-phenylphenyl)acetic acid | |
3332 | |
212.25 | |
4-Biphenylacetic acid |
158°C to 163°C | |
C14H12O2 | |
50 g | |
1211592 | |
4-biphenylacetic acid, felbinac, 4-biphenylylacetic acid, traxam, dolinac, 4-carboxymethylbiphenyl, napageln, 2-4-phenylphenyl acetic acid, p-biphenylylacetic acid, 1,1'-biphenyl-4-acetic acid | |
C1=CC=C(C=C1)C2=CC=C(C=C2)CC(=O)O | |
212.248 | |
CHEBI:31597 | |
98% |
Safety and Handling
GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H301-H315-H319-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 227-233-2
RTECSNumber : DU8229050
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only