missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Biphenylacetic acid, 98%

Manufacturer:  Alfa Aesar™ A1174918

 View more versions of this product

Catalog No. AAA1174918



Specifications

Specifications

4-Biphenylacetic acid
158°C to 163°C
5728-52-9
MFCD00004351
1211592
4-biphenylacetic acid, felbinac, 4-biphenylylacetic acid, traxam, dolinac, 4-carboxymethylbiphenyl, napageln, 2-4-phenylphenyl acetic acid, p-biphenylylacetic acid, 1,1'-biphenyl-4-acetic acid
C1=CC=C(C=C1)C2=CC=C(C=C2)CC(=O)O
212.248
CHEBI:31597
98%
Odorless
50g
C14H12O2
UN2811
14,3951
QRZAKQDHEVVFRX-UHFFFAOYSA-N
2-(4-phenylphenyl)acetic acid
3332
212.25

Chemical Identifiers

5728-52-9
212.248
QRZAKQDHEVVFRX-UHFFFAOYSA-N
3332
2-(4-phenylphenyl)acetic acid
C14H12O2
MFCD00004351
4-biphenylacetic acid, felbinac, 4-biphenylylacetic acid, traxam, dolinac, 4-carboxymethylbiphenyl, napageln, 2-4-phenylphenyl acetic acid, p-biphenylylacetic acid, 1,1'-biphenyl-4-acetic acid
CHEBI:31597
C1=CC=C(C=C1)C2=CC=C(C=C2)CC(=O)O
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : DU8229050

SDS Documents