Please login to your online account to display your discounted pricing

4-Benzylthiomorpholine 1,1-dioxide, 97%, Maybridge

Chemical Identifiers

Chemical Name or Material 4-Benzylthiomorpholine 1, 1-dioxide
CAS 26475-66-1
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C11H15NO2S
Formula Weight 225.31
MDL Number MFCD00084894
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

DFP00030EA

  SDS
fisher scientific company
DFP00030EA
10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 225.31
Quantity 10g
Assay 97%
CAS Max % 100.0
Chemical Name or Material 4-Benzylthiomorpholine 1, 1-dioxide
Assay Percent Range 97%
MDL Number MFCD00084894
CAS 26475-66-1
CAS Min % 96.5
Molecular Formula C11H15NO2S
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.