Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Benzylpyridine, 99%

$35.41 - $268.20

Chemical Identifiers

Chemical Name or Material 4-Benzylpyridine
CAS 2116-65-6
Molecular Formula C12H11N
Formula Weight 169.23
MDL Number MFCD00006443
Beilstein 115831
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1432618

 
alfa aesar™
A1432618
50g Each for $35.41

AAA1432622

 
alfa aesar™
A1432622
100g Each for $64.49

AAA1432636

 
alfa aesar™
A1432636
500g Each for $268.20
Description & Specifications

Specifications

Formula Weight 169.23
Molecular Formula C12H11N
RTECS Number US2625000
Flash Point 115°C (239°F)
Health Hazard 1 Danger
CAS 2116-65-6
Beilstein 115831
Quantity 50g
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 4-Benzylpyridine
TSCA TSCA
Boiling Point 286° to 287°C
MDL Number MFCD00006443
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 9° to 11°C
Assay 99%

Applications
4-Benzylpyridine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, dyes and agrochemicals.

Solubility
Soluble in ethanol, ethyl ether. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Incompatible with strong acids.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.