missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Benzylpyridine 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 106161000

 View more versions of this product

Catalog No. AC106161000


 

Specifications Description & Specifications

Specifications

≥96.0%
169.23
Clear slightly yellow
100.0
97%
C12H11N
2116-65-6
97%
1.5810 to 1.5830 (20°C, 589nm)
Liquid
100mL
20, 426
96.0
MFCD00006443
C6H5CH2(C5H4N)
4-Benzylpyridine
Authentic
Glass bottle

Chemical Identifiers

2116-65-6
100.0
C12H11N
169.23
4-Benzylpyridine, 97%
96.0
97%
C6H5CH2(C5H4N)
MFCD00006443
≥96.0%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 218-319-0

RTECSNumber : US2625000

TSCA : TSCA