Learn More
4-Benzylpyridine, 97%, ACROS Organics™
$37.39 - $46.75
Chemical Identifiers
CAS | 2116-65-6 |
---|---|
Molecular Formula | C12H11N |
Molecular Weight (g/mol) | 169.23 |
MDL Number | MFCD00006443 |
InChI Key | DBOLXXRVIFGDTI-UHFFFAOYSA-N |
Synonym | .gamma.-benzylpyridine, 1p29gqm0oh, 4-benzyl pyridine, 4-benzyl-pyridine, 4-benzylpyridine, 4-phenylmethyl pyridine, gamma-benzylpyridine, pyridine, 4-benzyl, pyridine, 4-phenylmethyl, unii-1p29gqm0oh |
PubChem CID | 16458 |
IUPAC Name | 4-benzylpyridine |
SMILES | C1=CC=C(C=C1)CC2=CC=NC=C2 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC106160050
|
Thermo Fisher Scientific
106160050 |
5mL | Glass bottle |
Each for $37.39
|
|
AC106161000
|
Thermo Fisher Scientific
106161000 |
100mL | Glass bottle |
Each for $46.75
|
|
Chemical Identifiers
2116-65-6 | |
169.23 | |
DBOLXXRVIFGDTI-UHFFFAOYSA-N | |
16458 | |
C1=CC=C(C=C1)CC2=CC=NC=C2 |
C12H11N | |
MFCD00006443 | |
.gamma.-benzylpyridine, 1p29gqm0oh, 4-benzyl pyridine, 4-benzyl-pyridine, 4-benzylpyridine, 4-phenylmethyl pyridine, gamma-benzylpyridine, pyridine, 4-benzyl, pyridine, 4-phenylmethyl, unii-1p29gqm0oh | |
4-benzylpyridine |
Specifications
1.0600g/mL | |
Authentic | |
1.5810 to 1.5830 | |
287.0°C | |
9.0°C to 11.0°C | |
2116-65-6 | |
100.0 | |
C12H11N | |
MFCD00006443 | |
.gamma.-benzylpyridine, 1p29gqm0oh, 4-benzyl pyridine, 4-benzyl-pyridine, 4-benzylpyridine, 4-phenylmethyl pyridine, gamma-benzylpyridine, pyridine, 4-benzyl, pyridine, 4-phenylmethyl, unii-1p29gqm0oh | |
DBOLXXRVIFGDTI-UHFFFAOYSA-N | |
4-benzylpyridine | |
16458 | |
Liquid | |
4-Benzylpyridine, 97% |
115°C | |
Glass bottle | |
1.06 | |
Yellow | |
5mL | |
96.0 | |
96% min. (GC) | |
C6H5CH2(C5H4N) | |
20, 426 | |
Solubility in water: insoluble in water. Other solubilities: soluble in organic solvents,soluble in ethanol and ether | |
C1=CC=C(C=C1)CC2=CC=NC=C2 | |
169.23 | |
169.23 | |
97% |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 218-319-0
RTECSNumber : US2625000
TSCA : TSCA