missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Benzylpiperidine, TCI America™

$53.47 - $296.39

Chemical Identifiers

CAS 31252-42-3
Molecular Formula C12H17N
Molecular Weight (g/mol) 175.275
MDL Number MFCD00006006
InChI Key ABGXADJDTPFFSZ-UHFFFAOYSA-N
Synonym piperidine, 4-phenylmethyl, 4-phenylmethyl piperidine, 4-benzyl-piperidine, piperidine, 4-benzyl, phenyl 4-piperidyl methane, alpha-4-piperidyl toluene, unii-jdf1t4667s, 4-benzyl piperidine, 4-benzylpiperdine, 4-benzylpiperidin
PubChem CID 31738
IUPAC Name 4-benzylpiperidine
SMILES C1CNCCC1CC2=CC=CC=C2
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B038725ML View Documents TCI America
B038725ML
25mL Each for $53.47
 
B0387250ML View Documents TCI America
B0387250ML
250mL Each for $296.39
 
Specifications

Chemical Identifiers

31252-42-3
175.275
ABGXADJDTPFFSZ-UHFFFAOYSA-N
31738
C1CNCCC1CC2=CC=CC=C2
C12H17N
MFCD00006006
piperidine, 4-phenylmethyl, 4-phenylmethyl piperidine, 4-benzyl-piperidine, piperidine, 4-benzyl, phenyl 4-piperidyl methane, alpha-4-piperidyl toluene, unii-jdf1t4667s, 4-benzyl piperidine, 4-benzylpiperdine, 4-benzylpiperidin
4-benzylpiperidine

Specifications

279°C
25mL
C12H17N
piperidine, 4-phenylmethyl, 4-phenylmethyl piperidine, 4-benzyl-piperidine, piperidine, 4-benzyl, phenyl 4-piperidyl methane, alpha-4-piperidyl toluene, unii-jdf1t4667s, 4-benzyl piperidine, 4-benzylpiperdine, 4-benzylpiperidin
C1CNCCC1CC2=CC=CC=C2
175.275
175.28
≥99.0% (GC)
Yellow
31252-42-3
MFCD00006006
ABGXADJDTPFFSZ-UHFFFAOYSA-N
4-benzylpiperidine
31738
Clear Liquid at 20°C
4-Benzylpiperidine
Safety and Handling

Safety and Handling

RTECSNumber : TM4728000

TSCA : Yes