missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Benzylpiperidine 98%, ACROS Organics™

Manufacturer:  Acros Organics 161931000

 View more versions of this product

Catalog No. AC161931000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

4-Benzylpiperidine
>112°C
Glass bottle
0.99
Colorless to Yellow
100g
31252-42-3
100.0
C12H17N
20, 296
ABGXADJDTPFFSZ-UHFFFAOYSA-N
4-benzylpiperidine
31738
Viscous Liquid
0.9900g/mL
Authentic
1.5370 to 1.5390
279.0°C
6.0°C to 7.0°C
98%
97.5
98%
MFCD00006006
piperidine, 4-phenylmethyl, 4-phenylmethyl piperidine, 4-benzyl-piperidine, piperidine, 4-benzyl, phenyl 4-piperidyl methane, alpha-4-piperidyl toluene, unii-jdf1t4667s, 4-benzyl piperidine, 4-benzylpiperdine, 4-benzylpiperidin
C1CNCCC1CC2=CC=CC=C2
175.275
175.27
98%

Chemical Identifiers

31252-42-3
175.275
ABGXADJDTPFFSZ-UHFFFAOYSA-N
31738
C1CNCCC1CC2=CC=CC=C2
C12H17N
MFCD00006006
piperidine, 4-phenylmethyl, 4-phenylmethyl piperidine, 4-benzyl-piperidine, piperidine, 4-benzyl, phenyl 4-piperidyl methane, alpha-4-piperidyl toluene, unii-jdf1t4667s, 4-benzyl piperidine, 4-benzylpiperdine, 4-benzylpiperidin
4-benzylpiperidine
Safety and Handling

Safety and Handling

EINECSNumber : 250-535-0

RTECSNumber : TM4728000

TSCA : TSCA

SDS Documents