missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Benzylpiperidine, 98%, ACROS Organics™
$18.00 - $18.00
Chemical Identifiers
CAS | 31252-42-3 |
---|---|
Molecular Formula | C12H17N |
Molecular Weight (g/mol) | 175.27 |
MDL Number | MFCD00006006 |
InChI Key | ABGXADJDTPFFSZ-UHFFFAOYSA-N |
Synonym | 4-benzyl piperidine, 4-benzyl-piperidine, 4-benzylpiperdine, 4-benzylpiperidin, 4-phenylmethyl piperidine, alpha-4-piperidyl toluene, phenyl 4-piperidyl methane, piperidine, 4-benzyl, piperidine, 4-phenylmethyl, unii-jdf1t4667s |
PubChem CID | 31738 |
IUPAC Name | 4-benzylpiperidine |
SMILES | C1CNCCC1CC2=CC=CC=C2 |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC161930050
|
Thermo Fisher Scientific
161930050 |
5g | Glass bottle |
Each for $18.00
|
|
AC161931000
|
Thermo Fisher Scientific
161931000 |
100g | Glass bottle |
N/A
|
N/A |
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
31252-42-3 | |
175.27 | |
ABGXADJDTPFFSZ-UHFFFAOYSA-N | |
31738 | |
C1CNCCC1CC2=CC=CC=C2 |
C12H17N | |
MFCD00006006 | |
4-benzyl piperidine, 4-benzyl-piperidine, 4-benzylpiperdine, 4-benzylpiperidin, 4-phenylmethyl piperidine, alpha-4-piperidyl toluene, phenyl 4-piperidyl methane, piperidine, 4-benzyl, piperidine, 4-phenylmethyl, unii-jdf1t4667s | |
4-benzylpiperidine |
Specifications
0.9900g/mL | |
Authentic | |
1.5370 to 1.5390 | |
279.0°C | |
6.0°C to 7.0°C | |
31252-42-3 | |
100.0 | |
C12H17N | |
20, 296 | |
ABGXADJDTPFFSZ-UHFFFAOYSA-N | |
4-benzylpiperidine | |
31738 | |
Viscous Liquid | |
4-Benzylpiperidine, 98% |
>112°C | |
Glass bottle | |
0.99 | |
Colorless to Yellow | |
5g | |
97.5 | |
98% | |
MFCD00006006 | |
4-benzyl piperidine, 4-benzyl-piperidine, 4-benzylpiperdine, 4-benzylpiperidin, 4-phenylmethyl piperidine, alpha-4-piperidyl toluene, phenyl 4-piperidyl methane, piperidine, 4-benzyl, piperidine, 4-phenylmethyl, unii-jdf1t4667s | |
C1CNCCC1CC2=CC=CC=C2 | |
175.27 | |
175.27 | |
98% |
Safety and Handling
EINECSNumber : 250-535-0
RTECSNumber : TM4728000
TSCA : TSCA