missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Benzylpiperidine, 98%

$54.15 - $616.30

Chemical Identifiers

CAS 31252-42-3
Molecular Formula C12H17N
Molecular Weight (g/mol) 175.275
MDL Number MFCD00006006
InChI Key ABGXADJDTPFFSZ-UHFFFAOYSA-N
Synonym 4-benzyl piperidine, 4-benzyl-piperidine, 4-benzylpiperdine, 4-benzylpiperidin, 4-phenylmethyl piperidine, alpha-4-piperidyl toluene, phenyl 4-piperidyl methane, piperidine, 4-benzyl, piperidine, 4-phenylmethyl, unii-jdf1t4667s
PubChem CID 31738
IUPAC Name 4-benzylpiperidine
SMILES C1CNCCC1CC2=CC=CC=C2
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2388314
View Documents
Alfa Aesar™
B2388314
25g
Each for $54.15
 
AAB2388322
View Documents
Alfa Aesar™
B2388322
100g
Each for $145.01
 
AAB2388336
View Documents
Alfa Aesar™
B2388336
500g
Each for $616.30
 
Description

Description

Specifications

Chemical Identifiers

31252-42-3
175.275
ABGXADJDTPFFSZ-UHFFFAOYSA-N
31738
C1CNCCC1CC2=CC=CC=C2
C12H17N
MFCD00006006
4-benzyl piperidine, 4-benzyl-piperidine, 4-benzylpiperdine, 4-benzylpiperidin, 4-phenylmethyl piperidine, alpha-4-piperidyl toluene, phenyl 4-piperidyl methane, piperidine, 4-benzyl, piperidine, 4-phenylmethyl, unii-jdf1t4667s
4-benzylpiperidine

Specifications

0.997
1.537
6°C to 7°C
31252-42-3
MFCD00006006
4-benzyl piperidine, 4-benzyl-piperidine, 4-benzylpiperdine, 4-benzylpiperidin, 4-phenylmethyl piperidine, alpha-4-piperidyl toluene, phenyl 4-piperidyl methane, piperidine, 4-benzyl, piperidine, 4-phenylmethyl, unii-jdf1t4667s
C1CNCCC1CC2=CC=CC=C2
175.275
175.28
4-Benzylpiperidine
147°C (296°F)
278°C to 279°C
25g
C12H17N
132339
ABGXADJDTPFFSZ-UHFFFAOYSA-N
4-benzylpiperidine
31738
98%
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

EINECSNumber : 250-535-0

RTECSNumber : TM4728000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS