Learn More
4-Benzylpiperidine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2388336
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Benzylpiperidine | |
6°C to 7°C | |
278°C to 279°C | |
C12H17N | |
MFCD00006006 | |
132339 | |
ABGXADJDTPFFSZ-UHFFFAOYSA-N | |
4-benzylpiperidine | |
31738 | |
98% |
31252-42-3 | |
0.997 | |
147°C (296°F) | |
1.537 | |
500 g | |
piperidine, 4-phenylmethyl, 4-phenylmethyl piperidine, 4-benzyl-piperidine, piperidine, 4-benzyl, phenyl 4-piperidyl methane, alpha-4-piperidyl toluene, unii-jdf1t4667s, 4-benzyl piperidine, 4-benzylpiperdine, 4-benzylpiperidin | |
C(C1CCNCC1)C1=CC=CC=C1 | |
175.28 | |
175.28 |
Chemical Identifiers
31252-42-3 | |
175.28 | |
ABGXADJDTPFFSZ-UHFFFAOYSA-N | |
31738 | |
C(C1CCNCC1)C1=CC=CC=C1 |
C12H17N | |
MFCD00006006 | |
piperidine, 4-phenylmethyl, 4-phenylmethyl piperidine, 4-benzyl-piperidine, piperidine, 4-benzyl, phenyl 4-piperidyl methane, alpha-4-piperidyl toluene, unii-jdf1t4667s, 4-benzyl piperidine, 4-benzylpiperdine, 4-benzylpiperidin | |
4-benzylpiperidine |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Warning
EINECSNumber : 250-535-0
RTECSNumber : TM4728000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only