Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Benzylphenylacetonitrile, 97%

Chemical Identifiers

Chemical Name or Material 4-Benzylphenylacetonitrile
CAS 101096-72-4
Molecular Formula C15H13N
Formula Weight 207.28
MDL Number MFCD00060310
UN Number UN3439
Beilstein 3258509
Synonym 4-Benzylbenzyl cyanide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1933203

  SDS
alfa aesar™
L1933203
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 207.28
CAS 101096-72-4
Health Hazard 3 GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 4-Benzylphenylacetonitrile
Assay 97%
Molecular Formula C15H13N
UN Number UN3439
Beilstein 3258509
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301
Toxic if swallowed.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
MDL Number MFCD00060310
Synonym 4-Benzylbenzyl cyanide
Boiling Point 145° to 150°C (0.05mmHg)
Melting Point 34° to 37°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301
Toxic if swallowed.