Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Benzylphenol, 98+%

$34.51 - $112.55

Chemical Identifiers

Chemical Name or Material 4-Benzylphenol
CAS 101-53-1
Molecular Formula C13H12O
Formula Weight 184.24
MDL Number MFCD00002384
Beilstein 1449572
Merck Index 14,1143
Synonym 4-Hydroxydiphenylmethane
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2253806

  SDS
alfa aesar™
B2253806
5g Each for $34.51

AAB2253814

  SDS
alfa aesar™
B2253814
25g Each for $112.55
Description & Specifications

Specifications

Formula Weight 184.24
Health Hazard 1 Warning
CAS 101-53-1
Merck Index 14,1143
Chemical Name or Material 4-Benzylphenol
Boiling Point 320° to 322°C
Quantity 5g
TSCA TSCA
Molecular Formula C13H12O
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Assay ≥98%
Synonym 4-Hydroxydiphenylmethane
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number SM8330500
Melting Point 81° to 85°C
Beilstein 1449572
MDL Number MFCD00002384
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.