Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Benzyloxypyridine N-oxide, 98%

$40.80 - $118.53

Chemical Identifiers

Chemical Name or Material 4-Benzyloxypyridine N-oxide
CAS 2683-66-1
Molecular Formula C12H11NO2
Formula Weight 201.22
MDL Number MFCD00047427
Beilstein 165005
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1794606

  SDS
alfa aesar™
L1794606
5g Each for $118.53

AAL1794603

  SDS
alfa aesar™
L1794603
1g Each for $40.80
Description & Specifications

Specifications

Formula Weight 201.22
Chemical Name or Material 4-Benzyloxypyridine N-oxide
Assay 98%
Molecular Formula C12H11NO2
CAS 2683-66-1
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
MDL Number MFCD00047427
Beilstein 165005
Melting Point 173° to 175°C
Quantity 5g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.