Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Benzyloxypropiophenone, 98%

$131.61 - $524.89

Chemical Identifiers

Chemical Name or Material 4'-Benzyloxypropiophenone
CAS 4495-66-3
Molecular Formula C16H16O2
Formula Weight 240.3
MDL Number MFCD00009311
Beilstein 2271608
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1832718

  SDS
alfa aesar™
A1832718
50g Each for $131.61

AAA1832730

  SDS
alfa aesar™
A1832730
250g Each for $524.89
Description & Specifications

Specifications

Formula Weight 240.3
Chemical Name or Material 4'-Benzyloxypropiophenone
Assay 98%
Molecular Formula C16H16O2
MDL Number MFCD00009311
CAS 4495-66-3
Beilstein 2271608
Melting Point 98° to 102°C
Quantity 50g