missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Benzyloxyphenylacetyl Chloride, TCI America™

$264.31 - $850.98

Chemical Identifiers

CAS 39188-62-0
Molecular Formula C15H13ClO2
Molecular Weight (g/mol) 260.717
MDL Number MFCD00674192
InChI Key GZAGJMNAVUCWGM-UHFFFAOYSA-N
PubChem CID 11357442
IUPAC Name 2-(4-phenylmethoxyphenyl)acetyl chloride
SMILES C1=CC=C(C=C1)COC2=CC=C(C=C2)CC(=O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B17805G View Documents TCI America
B17805G
5g Each for $264.31
 
B178025G View Documents TCI America
B178025G
25g Each for $850.98
 
Specifications

Chemical Identifiers

39188-62-0
260.717
GZAGJMNAVUCWGM-UHFFFAOYSA-N
2-(4-phenylmethoxyphenyl)acetyl chloride
C15H13ClO2
MFCD00674192
11357442
C1=CC=C(C=C1)COC2=CC=C(C=C2)CC(=O)Cl

Specifications

Undesignated
5g
C15H13ClO2
3261
C1=CC=C(C=C1)COC2=CC=C(C=C2)CC(=O)Cl
260.717
260.72
≥96.0% (T)
78°C
39188-62-0
MFCD00674192
GZAGJMNAVUCWGM-UHFFFAOYSA-N
2-(4-phenylmethoxyphenyl)acetyl chloride
11357442
Crystal-Powder at 20°C
4-Benzyloxyphenylacetyl Chloride
Safety and Handling

Safety and Handling

TSCA : No