Please login to your online account to display your discounted pricing

4-Benzyloxyphenylacetonitrile, +98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Benzyloxyphenylacetonitrile
CAS 838-96-0
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C15H13NO
Linear Formula C6H5CH2OC6H4CH2CN
Formula Weight 223.27
MDL Number MFCD00016400
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC394060050

  SDS
thermo fisher scientific
AC394060050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥98.0%
Physical Form Powder
Formula Weight 223.27
Quantity 5g
Color Cream
Melting Point 65° to 68°C
CAS Min % 98.0
CAS Max % 100.0
Linear Formula C6H5CH2OC6H4CH2CN
Chemical Name or Material 4-Benzyloxyphenylacetonitrile
MDL Number MFCD00016400
Infrared Spectrum Authentic
Packaging Glass bottle
CAS 838-96-0
Molecular Formula C15H13NO
Assay Percent Range 98+%
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.