missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Benzyloxyphenylacetonitrile, 97%

$41.00 - $363.51

Chemical Identifiers

CAS 838-96-0
Molecular Formula C15H13NO
Formula Weight 223.28
MDL Number MFCD00016400
Beilstein 2377584
Synonym 4-Benzyloxybenzyl cyanide
Chemical Name or Material 4-Benzyloxyphenylacetonitrile
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0977106 SDS Alfa Aesar™
L0977106
5g Each for $363.51
AAL09771MD SDS Alfa Aesar™
L09771MD
250mg Each for $41.00
AAL0977103 SDS Alfa Aesar™
L0977103
1g Each for $101.58
Specifications Description & Specifications

Chemical Identifiers

838-96-0
223.28
2377584
4-Benzyloxyphenylacetonitrile
C15H13NO
MFCD00016400
4-Benzyloxybenzyl cyanide

Specifications

838-96-0
223.28
2377584
170° to 173°C (0.1mmHg)
5g
97%
C15H13NO
MFCD00016400
4-Benzyloxybenzyl cyanide
66° to 70°C
4-Benzyloxyphenylacetonitrile

Safety and Handling

GHS H Statement
H302-H312
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P280h
Wear protective gloves/clothing.

Warning