Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Benzyloxyphenylacetic acid, 99%

$33.55 - $33.55

Chemical Identifiers

Chemical Name or Material 4-Benzyloxyphenylacetic acid
CAS 6547-53-1
Molecular Formula C15H14O3
Formula Weight 242.28
MDL Number MFCD00017540
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5090203

  SDS
alfa aesar™
H5090203
1g Each for $33.55

AAH5090206

  SDS
alfa aesar™
H5090206
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5090214

  SDS
alfa aesar™
H5090214
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 242.28
Chemical Name or Material 4-Benzyloxyphenylacetic acid
Assay 99%
Molecular Formula C15H14O3
MDL Number MFCD00017540
CAS 6547-53-1
Melting Point 119° to 121°C
Quantity 1g
Recommended Storage 2° to 8°C
Safety and Handling

2° to 8°C