missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Benzyloxyphenyl isothiocyanate, 98%

Manufacturer:  Alfa Aesar™ L1003803

 View more versions of this product

Catalog No. AAL1003803



Description

Description

Solubility
Hydrolyzes in water.

Notes
Air and moisture sensitive. Store at room temperature. Incompatible with strong oxidizing agents.
Specifications

Specifications

4-Benzyloxyphenyl isothiocyanate
1g
C14H11NOS
UN2923
4-benzyloxyphenyl isothiocyanate, 4-benzyloxyphenylisothiocyanate, 1-benzyloxy-4-isothiocyanatobenzene, 4-phenylmethoxy benzenisothiocyanate, acmc-20amoj, 4-benzyloxyisothiocyanate, 4-phenylmethoxyphenylisothiocyanate, 4-benzyloxy phenyl isothiocyanate, 4-phenylmethoxy phenyl isothiocyanate, 1-isothiocyanato-4-phenylmethoxy-benzene
C1=CC=C(C=C1)COC2=CC=C(C=C2)N=C=S
241.308
241.31
62°C to 65°C
139768-71-1
MFCD00041095
3298091
OQXRBXAFSXVCCO-UHFFFAOYSA-N
1-isothiocyanato-4-phenylmethoxybenzene
2735503
98%

Chemical Identifiers

139768-71-1
241.308
OQXRBXAFSXVCCO-UHFFFAOYSA-N
2735503
C1=CC=C(C=C1)COC2=CC=C(C=C2)N=C=S
C14H11NOS
MFCD00041095
4-benzyloxyphenyl isothiocyanate, 4-benzyloxyphenylisothiocyanate, 1-benzyloxy-4-isothiocyanatobenzene, 4-phenylmethoxy benzenisothiocyanate, acmc-20amoj, 4-benzyloxyisothiocyanate, 4-phenylmethoxyphenylisothiocyanate, 4-benzyloxy phenyl isothiocyanate, 4-phenylmethoxy phenyl isothiocyanate, 1-isothiocyanato-4-phenylmethoxy-benzene
1-isothiocyanato-4-phenylmethoxybenzene
Safety and Handling

Safety and Handling

GHS H Statement
H331-H314-H318-H302-H312
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.

Documents