Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Benzyloxyphenol, 98+%

$53.04 - $1,005.01

Chemical Identifiers

Chemical Name or Material 4-Benzyloxyphenol
CAS 103-16-2
Molecular Formula C13H12O2
Formula Weight 200.24
MDL Number MFCD00002333
Beilstein 1958305
Merck Index 14,6248
Synonym Hydroquinone monobenzyl ether; Monobenzone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA148430B

  SDS
alfa aesar™
A148430B
1000g Each for $1,005.01

AAA1484314

  SDS
alfa aesar™
A1484314
25g Each for $53.04

AAA1484322

  SDS
alfa aesar™
A1484322
100g Each for $148.86
Description & Specifications

Specifications

Formula Weight 200.24
Synonym Hydroquinone Monobenzyl Ether; Monobenzone
Health Hazard 2 GHS H Statement
H319-H317
Causes serious eye irritation.
May cause an allergic skin reaction.
Quantity 1000g
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P363-P321-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wash contaminated clothing before reuse. Specific treatment (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.
Merck Index 14,6248
Assay ≥98%
Melting Point 118° to 122°C
Health Hazard 1 Warning
TSCA TSCA
Beilstein 1958305
Chemical Name or Material 4-Benzyloxyphenol
MDL Number MFCD00002333
RTECS Number SJ7700000
Molecular Formula C13H12O2
CAS 103-16-2

Applications
4-Benzyloxyphenol is used in the synthesis of bis(4-benzyloxyphenoxy)phenyl phosphine oxide. It plays an essential role in the preparation of hetaryl-azophenol dyes via heterocyclic amines in nitrosyl sulphuric acid. It is also used for polyester fiber dyeing and in rubber industry. It acts as a depigmenting agent.

Solubility
Slightly soluble in water. Soluble in alcohol, acetone, benzene, and diethyl ether.

Notes
Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents and strong bases.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P363-P321-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wash contaminated clothing before reuse. Specific treatment (see label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H319-H317
Causes serious eye irritation.
May cause an allergic skin reaction.